Learn More
Ethos Biosciences Hematoxylin Powder Certified
Catalog No. 5000819177
Encompass_Preferred
Hematoxylin Powder is a light brownish-yellow crystalline powder derived from the heartwood of the logwood tree It is a natural phenolic compound that upon oxidation to hematein serves as the primary staining component in hematoxylin and other histological stains The powder is slighly soluble in water and ethanol and should be stored in a tighly sealed container protected from light and moisture
Catalog No. 50-008-19177 Supplier Ethos Biosciences Supplier No. 7575100
Hematoxylin Powder is a light brownish-yellow crystalline powder derived from the heartwood of the logwood tree It is a natural phenolic compound that upon oxidation to hematein serves as the primary staining component in hematoxylin and other histological stains The powder is slighly soluble in water and ethanol and should be stored in a tighly sealed container protected from light and moisture