Learn More
Scigen Hematoxylin Premium Stain
Catalog No. 23730631 Shop All Scigen Inc Products
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
23-730-631 1 L
1 options
This item is not returnable. View return policy
FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE.
Catalog No. 23-730-631 Supplier Scigen Supplier No. 7110
Add to Cart
Add to Cart
This item is not returnable. View return policy
FOR IN VITRO LABORATORY USE POISON: NOT FOR DRUG, FOOD, OR HOUSEHOLD USE.
A non-precipitating, progressive, mercury-free stain formulated to exceed the performance of Gills and Harris Hematoxylins.
Scigen™ Premium Hematoxylin (7110) is a non-precipitating, progressive, mercury-free stain formulated to exceed the performance of Gills and Harris Hematoxylins. It is an excellent nuclear stain for routine hematoxylin and eosin, frozen section, and IHC background staining.
Scigen™'s Hematoxylin produces incredible nuclear detail, with nuclear chromatin staining a crisp blue-purple with exceptional characteristics and provides the flexibility of Harris and Gills stains without staining acid mucopolysaccharides (mucin).
  • Made in the U.S.A.
  • Manufactured using ISO:13485 standard
Specifications
Certifications/Compliance Manufactured using ISO:13485 standard
For Use With (Application) Routine Hematoxylin and Eosin Staining, Frozen Section Stains and IHC
Quantity 1 L
Product Type Hematoxylin Stain