Learn More
Hematoxylin, Sigma-Aldrich
Catalog No. 01259055 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
01-259-055 25 g
1 options
Catalog No. 01-259-055 Supplier MilliporeSigma Supelco Supplier No. H313625G
Add to Cart
Add to Cart
Biological Stain Commission-certified hematoxylin for use in histological staining techniques including Weigert's, Mayer's, and Harris hemalum.
Certified for use in Weigert's iron hematoxylin, Mayer's hemalum, Harris hemalum and in Heidenhain's technique
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type Hematoxylin