Learn More
Sigma Aldrich Fine Chemicals Biosciences Hematoxylin Solution, Gill No. 3, MFCD00283324, 500 mL
Catalog No. NC0262362
Encompass
Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.. Used with hematoxylin and eosin staining.
Catalog No. NC0262362 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GHS316500ML
Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.. Used with hematoxylin and eosin staining.