Learn More
Sigma Aldrich Fine Chemicals Biosciences Hematoxylin Solution Mayer
Catalog No. 502904390
Encompass
Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.
Catalog No. 50-290-4390 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MHS1284L
Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.