Learn More
Sigma Aldrich Fine Chemicals Biosciences Hematoxylin Solution Mayer1L
Catalog No. 502901532
Encompass
Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.
Catalog No. 50-290-1532 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MHS321L
Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.