Learn More
Abcam Hemato x ylin Solution (Mayer's, Modified), 125ML
Catalog No. 502587710
Encompass_Preferred
Hematoxylin solution (Mayer's, Modified) is a high-quality, ready-to-use formulation of Modified Mayer's hematoxylin (Lille's modification), commonly used in histological staining. - Cited in over 10 publications - Nuclear stain – excellent for staining cell nuclei in tissue sections - Ready-to-use – no preparation required for immediate use - Commonly used in H&E staining – often used with eosin in Hematoxylin and Eosin (H&E) staining
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-7710 Supplier Abcam Supplier No. AB220365125ML
Hematoxylin solution (Mayer's, Modified) is a high-quality, ready-to-use formulation of Modified Mayer's hematoxylin (Lille's modification), commonly used in histological staining. - Cited in over 10 publications - Nuclear stain – excellent for staining cell nuclei in tissue sections - Ready-to-use – no preparation required for immediate use - Commonly used in H&E staining – often used with eosin in Hematoxylin and Eosin (H&E) staining
The product is subject to the following: Abcam Restricted Use Statement