Learn More
Newcomer Supply Hematoxylin Stain, Harris, 1 gallon. Ready to use, doesn’t require filtering and is mercury free.
SDP
Catalog No. NC1670663
Encompass
Newcomer Supply Hematoxylin Stain, Harris is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and does not contain glacial acetic acid or ethylene glycol.   This Harris formulation is designed for either manual or automated staining. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC1670663 Supplier Newcomer Supply Supplier No. 12013C
Newcomer Supply Hematoxylin Stain, Harris is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and does not contain glacial acetic acid or ethylene glycol.   This Harris formulation is designed for either manual or automated staining. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.