Learn More
Newcomer Supply Hematoxylin Stain, Harris Modified, 1 L. Ready to use, doesn’t require filtering, is enhanced with glacial acetic acid along with ethylene glycol and is mercury free.
SDP
Catalog No. NC2101902
Encompass
Newcomer Supply Hematoxylin Stain, Harris Modified is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and can be used in either manual or automated staining platforms. This modified Harris formulation contains glacial acetic acid for more precise nuclear staining and ethylene glycol to increase solution stability and reduce surface precipitate. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101902 Supplier Newcomer Supply Supplier No. 1201B
Newcomer Supply Hematoxylin Stain, Harris Modified is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and can be used in either manual or automated staining platforms. This modified Harris formulation contains glacial acetic acid for more precise nuclear staining and ethylene glycol to increase solution stability and reduce surface precipitate. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.