Learn More
Newcomer Supply Hematoxylin Stain, Harris, 1 L. Ready to use, doesn’t require filtering and is mercury free.
SDP
Catalog No. NC2101901
Encompass
Newcomer Supply Hematoxylin Stain, Harris is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and does not contain glacial acetic acid or ethylene glycol.   This Harris formulation is designed for either manual or automated staining. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101901 Supplier Newcomer Supply Supplier No. 12013B
Newcomer Supply Hematoxylin Stain, Harris is a ready to use regressive hematoxylin that does not require filtering, is mercury-free and does not contain glacial acetic acid or ethylene glycol.   This Harris formulation is designed for either manual or automated staining. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.