Learn More
Newcomer Supply Hematoxylin Stain, Mayer Modified, 4 Liters. Progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol.
SDP
Catalog No. NC2016149
Encompass
Newcomer Supply Hematoxylin Stain, Mayer Modified is a ready to use progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol. Due to the progressive staining nature of Mayer hematoxylin, over-staining is less likely and a differentiation step is not required.  Mayer hematoxylin is an excellent choice as a counterstain in immunohistochemistry (IHC), amyloid and copper staining procedures. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2016149 Supplier Newcomer Supply Supplier No. 1202C
Newcomer Supply Hematoxylin Stain, Mayer Modified is a ready to use progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol. Due to the progressive staining nature of Mayer hematoxylin, over-staining is less likely and a differentiation step is not required.  Mayer hematoxylin is an excellent choice as a counterstain in immunohistochemistry (IHC), amyloid and copper staining procedures. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.