Learn More
Newcomer Supply Hematoxylin Stain Set, Weigert Iron, 1L set - Excellent for Mucin & Trichrome Stains
SDP
Catalog No. NC2101941
Encompass
Solution A: Ferric Chloide, Acidified 500 ml, Solution B: Hematoxylin 1%, Alcoholic 500 ml. Newcomer Supply Hematoxylin Stain Set, Weigert Iron is the preferred nuclear stain in conjunction with trichrome and mucin stains. Iron hematoxylin is the optimal nuclear stain when succeeding stains are lengthy or acidic, where the use of an aluminum-mordanted hematoxylin stain would have a tendency to decolorize. Shelf Life is 2 years from date of manufacture. Store at 15-30°C
This item is not returnable. View return policy
Catalog No. NC2101941 Supplier Newcomer Supply Supplier No. 1409A
Solution A: Ferric Chloide, Acidified 500 ml, Solution B: Hematoxylin 1%, Alcoholic 500 ml. Newcomer Supply Hematoxylin Stain Set, Weigert Iron is the preferred nuclear stain in conjunction with trichrome and mucin stains. Iron hematoxylin is the optimal nuclear stain when succeeding stains are lengthy or acidic, where the use of an aluminum-mordanted hematoxylin stain would have a tendency to decolorize. Shelf Life is 2 years from date of manufacture. Store at 15-30°C