Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematoxylin Stain Solution, Gill 1 Formulation, Regular Strength, Ricca Chemical
SDP

Catalog No. 35355 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3535-5 20 L Cubitainer
3535-16 500 mL Natural Poly Bottle
3535-32 1 L Natural Poly Bottle
3535-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3535-5 Supplier Ricca Chemical Company Supplier No. 35355
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Staining Delicate Nuclei in Progressive Cytology Staining Procedures

Specifications

Chemical Name or Material Hematoxylin Stain Solution
Name Note Gill 1 Formulation, Regular Strength
CAS 7732-18-5, 107-21-1, 64-19-7, 7784-31-8, 517-28-2, 7681-55-2
CAS Min % 74.0, 20.68, 1.56, 1.31, 0.15, 0.01
CAS Max % 77.0, 21.52, 1.62, 1.36, 0.15, 0.02
Packaging Cubitainer
Color Dark Purple
pH 2 to 4
Quantity 20 L
Physical Form Liquid
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.