Learn More
Newcomer Supply Hematoxylin Stain, Mayer Modified, 500 ml. Progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol.
SDP
Catalog No. NC0084777
Encompass
Newcomer Supply Hematoxylin Stain, Mayer Modified is a ready to use progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol. Due to the progressive staining nature of Mayer hematoxylin, over-staining is less likely and a differentiation step is not required.  Mayer hematoxylin is an excellent choice as a counterstain in immunohistochemistry (IHC), amyloid and copper staining procedures. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC0084777 Supplier Newcomer Supply Supplier No. 1202A
Newcomer Supply Hematoxylin Stain, Mayer Modified is a ready to use progressive hematoxylin that does not require filtering and does not contain chloral hydrate or alcohol. Due to the progressive staining nature of Mayer hematoxylin, over-staining is less likely and a differentiation step is not required.  Mayer hematoxylin is an excellent choice as a counterstain in immunohistochemistry (IHC), amyloid and copper staining procedures. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.