Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematoxylin Stain Solution, Modified Harris Formulation, Mercury Free Nuclear Stain, Ricca Chemical
SDP

Catalog No. 35305 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3530-5 20 L
3530-16 500 mL
3530-32 1 L
3530-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3530-5 Supplier Ricca Chemical Company Supplier No. 35305
Add to Cart
Add to Cart

For General Histology and Cytology

Specifications

Color Purple
Description Modified Harris Formulation (Mercury Free)
Form Liquid
Packaging Cubitainer
Purity Mercury Free
Quantity 20 L
Product Type Hematoxylin Stain Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.