Learn More
Sigma Aldrich Fine Chemicals Biosciences Heme Assay Kit sufficient
Catalog No. 502904259
Encompass
Heme is a member of the porphyrin family. It is synthesized in both mitochondria and cytoplasm and is a key prosthetic group for various essential proteins such as hemoglobin cytochromes catalases and peroxidases. free heme which can be released from hemoglobin following hemolysis is pro-inflammatory and contributes to iron-derived reactive oxygen species. Heme determination is widely practiced by researchers of various blood diseases.
Catalog No. 50-290-4259 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3161KT
Heme is a member of the porphyrin family. It is synthesized in both mitochondria and cytoplasm and is a key prosthetic group for various essential proteins such as hemoglobin cytochromes catalases and peroxidases. free heme which can be released from hemoglobin following hemolysis is pro-inflammatory and contributes to iron-derived reactive oxygen species. Heme determination is widely practiced by researchers of various blood diseases.