Learn More
ABclonal Technology Heme Oxygenase 1 (HO-1/HMOX1) Rabbit pAb
SDP
Supplier:  ABclonal Technology A13461000UL
Encompass_Preferred
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.
Catalog No. 50-001-15833