Learn More
Enzo Life Sciences Heme oxygenase 1 polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLHC30010025
Encompass_Preferred
The peptide is totally conserved in pig, rat and mouse HO-1; three amino acid substitutions (A16L; A20G; V24A) exist in the aligned region (residues 31-44) within human, rat and rabbit HO-2; and a single change (A16L) exists in chicken HO. Immunogen: Synthetic peptide corresponding to aa 12-25 of human heme oxygenase 1 (HO-1) protein. Source: Purified from rabbit serum. Species reactivity: Human. Applications: IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-1024