Learn More
Enzo Life Sciences Heme oxygenase 2 polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLHC30020025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 246-264 of human heme oxygenase 2 (HO-2) protein (E.C. 1.14.99.3). Source: Purified from rabbit serum. Species reactivity: Human, Rat. Applications: IHC, WB. Purity: Protein G-affinity purified.
Catalog No. 50-201-1026