Learn More
American Radiolabeled Chemicals Inc HEMICHOL 3H DASALT 250UC
SDP
Catalog No. 50901111
Encompass_Preferred
Hemicholinium-methyl-3H 3-diacetate salt 250uCi in ethanol, 1 mCi/ml, S.A. 100-160 Ci/mmol, M.W. 532, Shipped in Dry Ice
Catalog No. 50-901-111 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1772250UC
Hemicholinium-methyl-3H 3-diacetate salt 250uCi in ethanol, 1 mCi/ml, S.A. 100-160 Ci/mmol, M.W. 532, Shipped in Dry Ice