Learn More
TARGETMOL CHEMICALS INC HEMICHOLINIUM 3 25MG
SDP
Catalog No. 502580968
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hemicholinium 3 is a potent and selective choline uptake blocker. purity: 99%
Catalog No. 50-258-0968 Supplier TARGETMOL CHEMICALS INC Supplier No. T222725MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hemicholinium 3 is a potent and selective choline uptake blocker. purity: 99%