Learn More
Echelon Biosciences Research Labs HEMIN 5 g
SDP
Catalog No. 50720328
Encompass
Hemin induces the activity of the enzyme heme-oxygenase.Heme oxygenase catalyzes the conversion of heme to biliverdin, CO and Fe+3. The induction of heme oxygenase activity has been associated with reduced free radical formation and inflammation, vascular repair, and implicated in tumor growth and metastasis. Hemin has been shown to be effective at 75 mol/kg in mice or 5 M for cells in culture. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-328 Supplier Echelon Biosciences Research Labs Supplier No. FH0105G
Hemin induces the activity of the enzyme heme-oxygenase.Heme oxygenase catalyzes the conversion of heme to biliverdin, CO and Fe+3. The induction of heme oxygenase activity has been associated with reduced free radical formation and inflammation, vascular repair, and implicated in tumor growth and metastasis. Hemin has been shown to be effective at 75 mol/kg in mice or 5 M for cells in culture. This item is non returnable