Learn More
Cayman Chemical Hemin chloride, Purity: ≥96%, CAS: 16009-13-5, 10 g
SDP
Catalog No. 502972556
Encompass_Preferred
An oxidized form of heme that inhibits eIF2αK1, a repressor of eIF-2α; used experimentally to induce the expression of heme oxygenase-1 in cells and in animals
Catalog No. 50-297-2556 Supplier Cayman Chemical Supplier No. 1648710 G
An oxidized form of heme that inhibits eIF2αK1, a repressor of eIF-2α; used experimentally to induce the expression of heme oxygenase-1 in cells and in animals