Learn More
Crescent Chemical Co Inc HEMIN CRYST. RESEARCH GRADE
SDP
Catalog No. 50247705
Encompass_Preferred
Hemin cryst. research grade Highly purified standard substance; specially developed for determination of hemoglobin (1). As labelling catalyst for luminescence immunoassay (2). Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number -
Catalog No. 50-247-705 Supplier Crescent Chemical Co Inc Supplier No. SE24410.02
Hemin cryst. research grade Highly purified standard substance; specially developed for determination of hemoglobin (1). As labelling catalyst for luminescence immunoassay (2). Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number -