Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Hemispheric Bowl for 50 mL Flask
SDP

Catalog No. 11392110
Change view
Click to view available options
For Use With (Equipment):
AREX-6 Hotplate Stirrers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-392-110 AREX-6 Hotplate Stirrers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-110 Supplier Velp™ Supplier No. A00000330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with AREX-6 Hotplate Stirrers. Suitable solution to replace oil baths and mantles in order to perform clean and safe analysis with round bottom flasks.

  • Holds various sizes of flasks
  • For use with 135 mm plate hotplates

Specifications

Capacity (Metric) 50 mL
Description Fits on 135 mm plate
For Use With (Equipment) AREX-6 Hotplate Stirrers
Type Base Plate
Warranty 24 Months
Material Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.