Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0411268
Encompass_Preferred
Fabric hemispherical mantle for 250ml spherical flask,heats to 400 degrees C maximum internal operating temperature, 180W, 115V CSA
Catalog No. NC0411268 Supplier Glas Col LLC Supplier No. 100A O402
Fabric hemispherical mantle for 250ml spherical flask,heats to 400 degrees C maximum internal operating temperature, 180W, 115V CSA