Learn More
Cliawaived Inc Eurotrol Hemotrol Level 1 (Low Control)
SDP
Catalog No. NC1784273
Encompass
EuroTrol Hemotrol Level 1 (Low Control) The Eurotrol Hemotrol Controls are assayed for the HemoCue Hb 201 and HemoCue B-Hemoglobin systems. The EuroTrol HemoTrol hemolysate is an assayed control solution for evaluating the accuracy and precision of the system. The control material is available in three different levels: Low, Normal and High. Each Eurotrol Hemotrol control vial is stable one-month from date of opening. All un-opened controls are stable through expiration when stored refrigerated.
This item is not returnable. View return policy
Catalog No. NC1784273 Supplier Cliawaived Inc Supplier No. EUR171001002
EuroTrol Hemotrol Level 1 (Low Control) The Eurotrol Hemotrol Controls are assayed for the HemoCue Hb 201 and HemoCue B-Hemoglobin systems. The EuroTrol HemoTrol hemolysate is an assayed control solution for evaluating the accuracy and precision of the system. The control material is available in three different levels: Low, Normal and High. Each Eurotrol Hemotrol control vial is stable one-month from date of opening. All un-opened controls are stable through expiration when stored refrigerated.