Learn More
Utah Medical Products Inc HEMO-NATE 18UM FILTER 50/BX
SDP
Supplier:  Utah Medical Products Inc 4020009
Encompass
Hemo-Nate� 18 Micron Blood Filter. The blood filtration set is designed for reducing risk of pulmonary distress after the infusion of frequently administered small volumes of blood, blood components and other fluids in neonatal and pediatric patients. 50/BOX
Catalog No. NC0474850