Learn More
HemoCue America 201 Analyzer Printer
Catalog No. 22601030
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
22-601-030 Each
1 options
Catalog No. 22-601-030 Supplier HemoCue America Supplier No. 130662
Martel Printer
  • RS-232 (printer) interface cable required but not supplied (Cat. No. 13-907-2)
  • Printer also compatible with Albumin 201, Glucose 201, Hb 201 and Hb 301 analyzers
TRUSTED_SUSTAINABILITY
Specifications
Product Type 201 Analyzer Printer
For Use With (Equipment) Albumin 201, Glucose 201, Hb 201 and Hb 301 analyzer
Quantity Each