Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America Hb 201+ Analyzer: Cuvettes

Catalog No. 22601002
Change view
Click to view available options
Quantity:
100
200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-601-002 100
22-601-003 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-601-002 Supplier HemoCue America Supplier No. 111732
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the HemoCue™ Hb 201+ Analyzer (22-601-007)

Break-resistant, disposable cuvette collects the exact amount of blood and mixes the sample with the reagents automatically.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cuvettes for HB-201 Analyzer
Sample Size 10 μL
Operating Temperature Range (English) 59°F to 86°F
Operating Temperature Range (Metric) 15°C to 30°C
For Use With (Equipment) Hb 201 analyzer
Quantity 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.