Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America WBC Analyzer Controls

Catalog No. 22601019
Change view
Click to view available options
Quantity:
3 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-601-019 3 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-601-019 Supplier HemoCue America Supplier No. WBC00S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To monitor values obtained from HemoCue WBC analyzer

  • Tri-level control: Low, Normal, High
  • 105-days closed vial stability and 30-days open vial stability
  • Packaged in 2mL plastic dropper vials

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CLIA
Format Plastic Vial
For Use With (Equipment) To monitor values obtained from a HemoCue™ WBC analyzer
Type R and D WBC Control
Intended Use Determination of total WBC count
Level 1 Low, 1 Normal, 1 High
Quantity 3 x 2 mL
Stability 105 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.