Learn More
PixCell Medical™ HemoScreen HemoCup
Catalog No. 23123806
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
23-123-806 100 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-123-806 Supplier PixCell Medical™ Supplier No. ACMA00100
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
HemoCup is a disposable tool used to assist in dispensing venous blood samples from a K2EDTA tube.
  • HemoCup facilitates simple blood collection into the Test Kit Sampler for venous blood draw.
  • This easy to use tool is an alternative to dispensing K2EDTA blood samples with a pipette or directly filling the Sampler from a K2EDTA tube.
Specifications
Type Plastic Cup
Quantity 100 Pk.
Dimensions (L x W x H) 1.25 x 0.5 x 0.5 in.