Learn More
Weber Scientific Hemocytometer for Cell Counting with Bright Lines
SDP
Catalog No. NC1677130
Encompass
Hemocytometer for cell counting has an H-shaped moat form two counting areas, or plateaus and the “V” slash reduces the possibility of overflow. Cell Depth: 0.100mm +/- 2% (1/10mm); Volume: 0.1 Microliter; Ruling Pattern: Improved Neubauer, 1/400 Square mm
Catalog No. NC1677130 Supplier Weber Scientific Supplier No. 304811
Hemocytometer for cell counting has an H-shaped moat form two counting areas, or plateaus and the “V” slash reduces the possibility of overflow. Cell Depth: 0.100mm +/- 2% (1/10mm); Volume: 0.1 Microliter; Ruling Pattern: Improved Neubauer, 1/400 Square mm