Learn More
Medchemexpress LLC Hemoglobin | 9008-02-0 | MFCD00130638 | ≥98% | 64.45 kDa | 250 MG
SDP
Catalog No. 5000396205
Encompass_Preferred
Hemoglobin is an iron-containing oxygen-transport metalloprotein found in the red blood cells of almost all vertebrates. It carries oxygen from respiratory organs to the body and picks up carbon dioxide from tissues, transporting it back to respiratory organs. Hemoglobin also acts as an inducer of HO-1 and functions as an antioxidant by absorbing unneeded oxygen in tissues.
  • Iron-containing protein
  • Oxygen binding properties
  • Inducer of HO-1
  • Transports carbon dioxide, nitric oxide, hydrogen sulfide, and sulfide
  • Functions as an antioxidant
  • Suitable for in vitro biological studies
  • Provides protection against LPS-induced lethal endotoxemia in vivo
Catalog No. 50-003-96205 Supplier Medchemexpress LLC Supplier No. 250MGHYP2995
Hemoglobin is an iron-containing oxygen-transport metalloprotein found in the red blood cells of almost all vertebrates. It carries oxygen from respiratory organs to the body and picks up carbon dioxide from tissues, transporting it back to respiratory organs. Hemoglobin also acts as an inducer of HO-1 and functions as an antioxidant by absorbing unneeded oxygen in tissues.
  • Iron-containing protein
  • Oxygen binding properties
  • Inducer of HO-1
  • Transports carbon dioxide, nitric oxide, hydrogen sulfide, and sulfide
  • Functions as an antioxidant
  • Suitable for in vitro biological studies
  • Provides protection against LPS-induced lethal endotoxemia in vivo