Learn More
MyBioSource Hemoglobin A1c
SDP
Supplier:  MyBioSource MBS5376851 MG
Encompass_Preferred
Hemoglobin A1c is a glycated form of hemoglobin used primarily to identify the average plasma glucose concentration over prolonged periods of time. It is formed in a non-enzymatic glycation pathway by hemoglobin's exposure to plasma glucose. Normal levels of glucose produce a normal amount of glycated hemoglobin. As the average amount of plasma glucose increases, the fraction of glycated hemoglobin increases in a predictable way. This serves as a marker for average blood glucose levels over the previous months prior to the measurement.
Catalog No. NC1792872