Learn More
Santa Cruz Biotechnology HEMOGLOBIN B ANTBDY (37-8)FITC
Supplier:  Santa Cruz Biotechnology SC21757 FITC
Encompass
Hemoglobin β Antibody (37-8) FITC,200 µg/ml•Hemoglobin β Antibody (37-8) is a mouse monoclonal IgG1 (kappa light chain) provided at 200 µg/ml•raised against human Hemoglobin•recommended for detection of Hemoglobin β of human origin by WB, IP, IF, IHC(P) and FCM
Catalog No. NC1569707