Learn More
Cell Signaling Technology Hemoglobin Subunit Beta Monoclonal Antibody, Rabbit, Human, 100 μl
SDP
Catalog No. 502730745
Encompass
The Hemoglobin beta (D4W4I) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of the hemoglobin β subunit. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of the human hemoglobin β subunit. It is suitable for applications including Western blotting and flow cytometry (fixed/permeabilized).
  • Specifically recognizes endogenous hemoglobin β subunit
  • May also detect hemoglobin δ subunit
  • No predicted cross-reactivity with hemoglobin α, γ, ε or ζ subunits
  • Validated for Western blotting and flow cytometry (fixed/permeabilized) applications
  • Recombinant monoclonal rabbit IgG
Catalog No. 50-273-0745 Supplier Cell Signaling Technology Supplier No. 84934S
The Hemoglobin beta (D4W4I) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of the hemoglobin β subunit. This recombinant antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of the human hemoglobin β subunit. It is suitable for applications including Western blotting and flow cytometry (fixed/permeabilized).
  • Specifically recognizes endogenous hemoglobin β subunit
  • May also detect hemoglobin δ subunit
  • No predicted cross-reactivity with hemoglobin α, γ, ε or ζ subunits
  • Validated for Western blotting and flow cytometry (fixed/permeabilized) applications
  • Recombinant monoclonal rabbit IgG