Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Hemoglobin, Bovine

Catalog No. AAJ6383806
Change view
Click to view available options
Quantity:
5g
25g
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6383806 5g
AAJ6383814 25g
AAJ6383822 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6383806 Supplier Thermo Scientific Chemicals Supplier No. J6383806
Add to Cart

missing translation for 'validMainframeMsg'

For research use | 64.5KDa

Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. It is used as a coloring agent for pet foods, a natural source of iron for nutraceuticals, a protein source for non-ruminant animals, and as a raw material for pharmaceutical porphyrin derivative production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture - Iron Delivery
Formulation Lyophilized powder
Molecular Weight (g/mol) 64.5KDa
Quantity 5g
Source Bovine blood
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Hemoglobin, Bovine
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.