Learn More
Arbor Assays Hemoglobin Colorimetric Detection Kit (2 plate)
SDP
Supplier:  Arbor Assays K013H1
Encompass_Preferred
The DetectX Hemoglobin Colorimetric Detection Kit uses a single reaction solution that is stable at 4°C, is not light sensitive, and does not contain dangerous chemicals.
Catalog No. 50-227-8395