Learn More
Arbor Assays Hemoglobin High Sensitivity Colorimetric Detection Kit (10 Plate)
SDP
Catalog No. 502278397
Encompass
The DetectX Hemoglobin High Sensitivity Colorimetric Detection Kit uses a single reaction solution that is stable at 4°C, is not light sensitive, and does not contain dangerous chemicals.
This item is not returnable. View return policy
Catalog No. 50-227-8397 Supplier Arbor Assays Supplier No. K013HX5
The DetectX Hemoglobin High Sensitivity Colorimetric Detection Kit uses a single reaction solution that is stable at 4°C, is not light sensitive, and does not contain dangerous chemicals.