Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin Spray Dried Spray Dried MP Biomedicals

Catalog No. ICN15123590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15123590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15123590 Supplier MP Biomedicals, Inc Supplier No. 0215123590

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9008-02-0
Molecular Formula C13H10N2O2
Molecular Weight (g/mol) 226.24
MDL Number MFCD00131282
InChI Key INGWEZCOABYORO-UHFFFAOYSA-N
Synonym 7-methyl-2-2-furyl-1,8-naphthyridine-4 1h-one, d0y8up, 2-furan-2-yl-7-methyl-1h-1,8 naphthyridin-4-one
PubChem CID 13285535
IUPAC Name 2-(furan-2-yl)-7-methyl-1,4-dihydro-1,8-naphthyridin-4-one
SMILES CC1=CC=C2C(=O)C=C(NC2=N1)C1=CC=CO1

Specifications

Quantity 500 g
Formula Weight 64.5
Chemical Name or Material Hemoglobin Spray Dried, Spray Dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.