Learn More
Medchemexpress LLC Hemoglobin subunit alpha (HBA1) protein, human (N-6xHis tag) | >95.0% | 5UG
SDP
Catalog No. 5000175262
Encompass_Preferred
Hemoglobin subunit alpha (HBA1) is a recombinant human alpha-globin protein expressed in E. coli and supplied lyophilized with an N-terminal 6xHis tag. The preparation corresponds to the full alpha-globin sequence (residues M1-R142) and is provided with stated purity and storage guidance for use in biochemical assays, antibody validation, and structural or biophysical studies.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Full-length alpha-globin sequence (residues M1-R142).
  • N-terminal 6xHis tag for detection or affinity capture.
  • Expression in Escherichia coli for recombinant production.
  • Supplied lyophilized in buffered formulations for stability.
  • Available in small quantities suitable for assay development and validation.
Catalog No. 50-001-75262 Supplier Medchemexpress LLC Supplier No. HYP703975UG
Hemoglobin subunit alpha (HBA1) is a recombinant human alpha-globin protein expressed in E. coli and supplied lyophilized with an N-terminal 6xHis tag. The preparation corresponds to the full alpha-globin sequence (residues M1-R142) and is provided with stated purity and storage guidance for use in biochemical assays, antibody validation, and structural or biophysical studies.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Full-length alpha-globin sequence (residues M1-R142).
  • N-terminal 6xHis tag for detection or affinity capture.
  • Expression in Escherichia coli for recombinant production.
  • Supplied lyophilized in buffered formulations for stability.
  • Available in small quantities suitable for assay development and validation.