Learn More
Cayman Chemical Hemokinin 1 (human) (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502913238
Encompass_Preferred
A peptide agonist of NK1 receptors; induces migration of A375 and B16/F10 cells at 1 µM, an effect that can be reversed by L-732,138; promotes the proliferation, migration, and tube formation of HUVECs at 0.001-0.1 µM; induces the production of PGE2 in human colon mucosal and muscle explants; intracerebroventricular administration increases the latency to withdrawal in the tail-flick test in mice; decreases MAP and heart rate in anesthetized rats at 1-30 nmol/kg
Catalog No. 50-291-3238 Supplier Cayman Chemical Supplier No. 354585 MG
A peptide agonist of NK1 receptors; induces migration of A375 and B16/F10 cells at 1 µM, an effect that can be reversed by L-732,138; promotes the proliferation, migration, and tube formation of HUVECs at 0.001-0.1 µM; induces the production of PGE2 in human colon mucosal and muscle explants; intracerebroventricular administration increases the latency to withdrawal in the tail-flick test in mice; decreases MAP and heart rate in anesthetized rats at 1-30 nmol/kg