Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Hemolysis Wash Solution 80H
SDP

Catalog No. 21377291
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-377-291 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.
Catalog No. 21-377-291 Supplier Arkray Supplier No. 103398
Add to Cart
Add to Cart
Handle with care. Avoid contact with skin or eyes. Use in a well-ventilated area.

Arkray Hemolysis Wash Solution 80H efficiently removes hemolyzed samples, ensuring accurate and reliable results. Optimized for Arkray analyzers, it simplifies workflow and maintains consistent laboratory performance.

  • Provided as 2 L bottles, 3 bottles/box

Specifications

Sample Type Urine
Type Solution
Form Liquid

Arkray Hemolysis Wash Solution 80H efficiently clears hemolyzed samples, supporting accurate and consistent results while streamlining laboratory workflow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.