Learn More
Athens Research And Technology Inc HEMOPEXIN HU PLASMA 1MG
Supplier:  Athens Research And Technology Inc 16160805131MG
Encompass_Preferred
Hemopexin, Human Plasma: 1 MG; Lyophilized from 11.9 mM phosphate, 137 mM NaCl, 2.7 mM KCl, pH 7.4.; MW 57,000; EC=1.97; Purity:> =95% by SDS-PAGE. This native protein is prepared and purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-753