Learn More
EKF Diagnostics HemoPoint H2™ Hemoglobin Meter
SDP
Catalog No. 22022655 Shop All Stanbio Laboratory Products
Change view
Click to view available options
:
Hemoglobin Analyzer
Catalog No.
22-022-655 Hemoglobin Analyzer
1 options
This item is not returnable. View return policy
Catalog No. 22-022-655 Supplier EKF Diagnostics Supplier No. G3000CONTCUV
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View Alternate Product
This item is not returnable. View return policy
Hemoglobin Meter that provides hemoglobin measurement and hematocrit calculation
  • Clia waived
  • Touchscreen
  • 8 μ sample size
  • Recalls last 4,000 test results
  • Rechargeable battery
Specifications
Type Hemoglobin Analyzer