Learn More
EKF Diagnostics HemoPoint H2 Microcuvettes, 100 Count
SDP
Catalog No. 22022637 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
100
Catalog No. Quantity
22-022-637 100
1 options
This item is not returnable. View return policy
Catalog No. 22-022-637 Supplier EKF Diagnostics Supplier No. 3015100
Add to Cart
Add to Cart
This item is not returnable. View return policy
HemoPoint H2 Microcuvettes with low 8 μL sample size
  • Simple sampling from any angle, bubble-free technology, pre-filled with dry reagent, blood drawn in by capillary action
Specifications
Type Microcuvette
Quantity 100