Learn More
EKF Diagnostics HemoPoint H2 Microcuvettes, 200 Count
SDP
Catalog No. 22022638 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
200
Catalog No. Quantity
22-022-638 200
1 options
This item is not returnable. View return policy
Catalog No. 22-022-638 Supplier EKF Diagnostics Supplier No. 3015200
This item is not returnable. View return policy
HemoPoint H2 Microcuvettes with low 8 μL sample size
  • Simple sampling from any angle, bubble-free technology, pre-filled with dry reagent, blood drawn in by capillary action
Specifications
Type Microcuvette
Quantity 200