Learn More
Fine Science Tools Kelly Hemostats, Stainless Steel, Curved, 14 cm
SDP
Catalog No. NC9492472
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 22mm/ Alloy / Material: Stainless Steel/ Length: 14cm
Catalog No. NC9492472 Supplier Fine Science Tools Supplier No. 1301914
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 22mm/ Alloy / Material: Stainless Steel/ Length: 14cm