Learn More
Roboz Surgical HEMOSTATIC FORCEPS
SDP
Catalog No. NC9855232
Encompass
Halstead Mosquito Forceps; Extra Delicate; Straight; 5 in. length, 0.6mm tip diameter, 1.7mm jaw width These are our extra delicate Halstead Mosquito hemostats. The tip diameter is 0.6mm. The jaw width is 1.7mm at 5mm from the tip.
Catalog No. NC9855232 Supplier Roboz Surgical Supplier No. RS7112
Halstead Mosquito Forceps; Extra Delicate; Straight; 5 in. length, 0.6mm tip diameter, 1.7mm jaw width These are our extra delicate Halstead Mosquito hemostats. The tip diameter is 0.6mm. The jaw width is 1.7mm at 5mm from the tip.